missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CX3CL1/Fractalkine Full Length Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20312
Molecular Weight 34 kDa
Name CX3CL1/Fractalkine Full Length
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
472FF025 R&D Systems
RDS472FF025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
472FF025CF R&D Systems
RDS472FF025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Gln25-Arg337 , with a C-terminal 6-His tag
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 34 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 20312
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
34 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
20312
CX3CL1/Fractalkine Full Length
25μg
Mouse