missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CTLA-4 Fc Chimera Recombinant Protein

$481.01 - $481.01

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 12477
Molecular Weight 41 kDa (monomer)
Name CTLA-4 Fc Chimera
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
434CT200 R&D Systems
RDS434CT200
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $481.01
434CT200CF R&D Systems
RDS434CT200CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $481.01
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ala37-Phe162
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 41 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 12477
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
41 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
12477
CTLA-4 Fc Chimera
200μg
Mouse