missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Crossveinless-2/CV-2 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 73230
Name Crossveinless-2/CV-2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 50μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2299CV050 R&D Systems
RDS2299CV050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
2299CV050CF R&D Systems
RDS2299CV050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Val34-Arg685, with an N-terminal 9-His tag, Ala39-Asp369 & Pro370-Arg685
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 73230
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
Crossveinless-2/CV-2
50μg
Mouse
73230
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes