Please login to your online account to display your discounted pricing

R&D Systems Mouse COCO Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 23863
Molecular Weight 17.3 kDa (monomer)
Name COCO
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

3356CC025

 
r&d systems
RDS3356CC025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $419.81

3356CC025CF

 
r&d systems
RDS3356CC025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 23863
Molecular Weight 17.3 kDa (monomer)
Name COCO
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse

Source: E. coli-derived, Arg24-Leu185
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.3 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 23863
Regulatory Status: RUO