missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CMG-2/ANTXR2 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 71914
Molecular Weight 32 kDa
Name CMG-2/ANTXR2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3636CM050 R&D Systems
RDS3636CM050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
3636CM050CF R&D Systems
RDS3636CM050CF
Supplied as a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gly26-Gly318, with a C-terminal 6-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 32 kDa
Formulation: Supplied as a 0.2μm filtered solution in PBS.
Entrez Gene ID: 71914
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
32 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
71914
CMG-2/ANTXR2
50μg
Mouse