missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Chordin Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 12667
Molecular Weight 101.5 kDa
Name Chordin
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
758CN050CF R&D Systems
RDS758CN050CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
758CN050 R&D Systems
RDS758CN050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Thr27-Ser948
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 101.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 12667
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
101.5 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
12667
Chordin
50μg
Mouse