missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Cerberus 1 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 12622
Molecular Weight 28.7 kDa (monomer)
Name Cerberus 1
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1986CB025CF R&D Systems
RDS1986CB025CF
Lyophilized from a 0.2μm filtered solution in HCl. Each for $416.79
1986CB025 R&D Systems
RDS1986CB025
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. Each for $416.79
Specifications

Description

Source: E. coli-derived, Asp18-Pro272, with an N-terminal Met
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 28.7 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in HCl.
Entrez Gene ID: 12622
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
28.7 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
12622
Cerberus 1
25μg
Mouse