missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CD28 Fc Chimera Recombinant Protein

$467.46 - $467.46

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 12487
Molecular Weight 52 kDa (monomer)
Name CD28 Fc Chimera
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
483CD200 R&D Systems
RDS483CD200
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $467.46
483CD200CF R&D Systems
RDS483CD200CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $467.46
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Lys21-Lys149
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 52 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 12487
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
52 kDa (monomer)
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
12487
CD28 Fc Chimera
200μg
Mouse