missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CCL9/10/MIP-1 gamma Recombinant Protein

$378.79 - $378.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20308
Molecular Weight 11.5 kDa
Name CCL9/10/MIP-1 gamma
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
463MG025 R&D Systems
RDS463MG025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $378.79
463MG025CF R&D Systems
RDS463MG025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $378.79
Specifications

Description

Source: E. coli-derived, Gln22-Gln122
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 11.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 20308
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
11.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
20308
CCL9/10/MIP-1 gamma
25μg
Mouse