missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CCL7/MCP-3/MARC Recombinant Protein

$337.62 - $559.23

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20306
Molecular Weight 8.5 kDa
Name CCL7/MCP-3/MARC
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
456MC050 R&D Systems
RDS456MC050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 50μg Each for $559.23
456MC050CF R&D Systems
RDS456MC050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 50μg Each for $559.23
456MC010 R&D Systems
RDS456MC010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $337.62
456MC010CF R&D Systems
RDS456MC010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $337.62
Specifications

Description

Source: E. coli-derived, Gln24-Pro97
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 20306
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
20306
CCL7/MCP-3/MARC
Yes