missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CCL6/C10 Truncated, aa 42-116 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20305
Molecular Weight 8.3 kDa
Name CCL6/C10 Truncated, aa 42-116
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3210C025 R&D Systems
RDS3210C025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $416.79
3210C025CF R&D Systems
RDS3210C025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $416.79
Specifications

Description

Source: E. coli-derived, Gly42-Ala116
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.3 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 20305
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8.3 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
20305
CCL6/C10 Truncated, aa 42-116
25μg
Mouse