Please login to your online account to display your discounted pricing

R&D Systems Mouse CCL24/Eotaxin-2/MPIF-2 aa 27-119 Recombinant Protein


Manufacturer: r&d systems  RDS1017MP025

 View more versions of this product

Catalog No. 1017MP025

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 10.3 kDa
Name CCL24/Eotaxin-2/MPIF-2 aa 27-119
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse
Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 56221