Please login to your online account to display your discounted pricing

R&D Systems Mouse CCL24/Eotaxin-2/MPIF-2 aa 24-119 Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 56221
Molecular Weight 10.6 kDa
Name CCL24/Eotaxin-2/MPIF-2 aa 24-119
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

528MP025

 
r&d systems
RDS528MP025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $298.51

528MP025CF

 
r&d systems
RDS528MP025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $298.51
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 56221
Molecular Weight 10.6 kDa
Name CCL24/Eotaxin-2/MPIF-2 aa 24-119
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse

Source: E. coli-derived, Ile24-Val119
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 10.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 56221
Regulatory Status: RUO