Please login to your online account to display your discounted pricing

R&D Systems Mouse CCL22/MDC Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20299
Molecular Weight 7.8 kDa
Name CCL22/MDC
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

439MD025

 
r&d systems
RDS439MD025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $415.83

439MD025CF

 
r&d systems
RDS439MD025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $415.83
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20299
Molecular Weight 7.8 kDa
Name CCL22/MDC
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse

Source: E. coli-derived, Gly25-Ser92
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.8 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 20299
Regulatory Status: RUO