missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CCL21/6Ckine Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 18829
Molecular Weight 12 kDa
Name CCL21/6Ckine
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
4576C025CF R&D Systems
RDS4576C025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $415.83
4576C025 R&D Systems
RDS4576C025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $415.83
Specifications

Description

Source: E. coli-derived, Ser24-Gly133
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 18829
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
12 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
18829
CCL21/6Ckine
25μg
Mouse