missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CCL20/MIP-3 alpha Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20297
Molecular Weight 7.9 kDa
Name CCL20/MIP-3 alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
760M3025 R&D Systems
RDS760M3025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
760M3025CF R&D Systems
RDS760M3025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: E. coli-derived, Ala27-Met96
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 20297
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
20297
CCL20/MIP-3 alpha
25μg
Mouse