missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CCL2/JE/MCP-1 Recombinant Protein

$272.44 - $559.23

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20296
Molecular Weight 8.5 kDa
Name CCL2/JE/MCP-1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
479JE010 R&D Systems
RDS479JE010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10μg Each for $272.44
479JE010CF R&D Systems
RDS479JE010CF
Lyophilized from a 0.2μm filtered solution in PBS. 10μg Each for $272.44
479JE050 R&D Systems
RDS479JE050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50μg Each for $559.23
479JE050CF R&D Systems
RDS479JE050CF
Lyophilized from a 0.2μm filtered solution in PBS. 50μg Each for $559.23
Specifications

Description

Source: E. coli-derived, Gln24-Arg96
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 20296
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
20296
CCL2/JE/MCP-1
Yes