missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CCL19/MIP-3 beta Recombinant Protein

$290.11 - $290.11

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 24047
Molecular Weight 9.4 kDa
Name CCL19/MIP-3 beta
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
440M3025 R&D Systems
RDS440M3025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $290.11
440M3025CF R&D Systems
RDS440M3025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $290.11
Specifications

Description

Source: E. coli-derived, Gly26-Val107-Leu-Glu, with a substitution of Ser108LeuGlu
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 24047
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
9.4 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
24047
CCL19/MIP-3 beta
25μg
Mouse