missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CCL17/TARC Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20295
Molecular Weight 7.9 kDa
Name CCL17/TARC
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
529TR025 R&D Systems
RDS529TR025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $298.51
529TR025CF R&D Systems
RDS529TR025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $298.51
Specifications

Description

Source: E. coli-derived, Ala24-Pro93
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 7.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 20295
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
20295
CCL17/TARC
25μg
Mouse