missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse CCL11/Eotaxin Recombinant Protein

$290.11 - $860.18

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 20292
Molecular Weight 8.4 kDa
Name CCL11/Eotaxin
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
420ME020CF R&D Systems
RDS420ME020CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 20μg Each for $290.11
420ME100 R&D Systems
RDS420ME100
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 100μg Each for $860.18
420ME020 R&D Systems
RDS420ME020
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 20μg Each for $290.11
420ME100CF R&D Systems
RDS420ME100CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 100μg Each for $860.18
Specifications

Description

Source: E. coli-derived, His24-Pro97
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 20292
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
8.4 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
20292
CCL11/Eotaxin
Yes