missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Cardiotrophin-1/CT-1 Recombinant Protein

$350.65 - $1,127.58

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 13019
Molecular Weight 21 kDa
Name Cardiotrophin-1/CT-1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
438CT010 R&D Systems
RDS438CT010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $350.65
438CT010CF R&D Systems
RDS438CT010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 10μg Each for $350.65
438CT050CF R&D Systems
RDS438CT050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 50μg Each for $1,127.58
438CT050 R&D Systems
RDS438CT050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 50μg Each for $1,127.58
Specifications

Description

Source: E. coli-derived, Ser2-Ala203
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 13019
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
21 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mouse
13019
Cardiotrophin-1/CT-1
Yes