Please login to your online account to display your discounted pricing

R&D Systems Mouse BMPR-IB/ALK-6 Fc Chimera Recombinant Protein

$521.89 - $521.89

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 12167
Molecular Weight 40.6 kDa (monomer)
Name BMPR-IB/ALK-6 Fc Chimera
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

444BNS100

 
r&d systems
RDS444BNS100
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $521.89

444BNS100CF

 
r&d systems
RDS444BNS100CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $521.89
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 12167
Molecular Weight 40.6 kDa (monomer)
Name BMPR-IB/ALK-6 Fc Chimera
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 100μg
Recombinant Yes
Source Mouse

Source: Mouse myeloma cell line, NS0-derived, Leu2-Lys126
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 40.6 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 12167
Regulatory Status: RUO