missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse BMP-7 Recombinant Protein

$533.15 - $533.15

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 12162
Molecular Weight 15.6 kDa (monomer)
Name BMP-7
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
5666BP010CF R&D Systems
RDS5666BP010CF
Lyophilized from a 0.2μm filtered solution in HCl. Each for $533.15
5666BP010 R&D Systems
RDS5666BP010
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. Each for $533.15
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Ser292-His430
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.6 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in HCl.
Entrez Gene ID: 12162
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
15.6 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
12162
BMP-7
10μg
Mouse