missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse BMP-4 Recombinant Protein

$572.26 - $572.26

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 12159
Molecular Weight 13.1 kDa (monomer)
Name BMP-4
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
5020BP010 R&D Systems
RDS5020BP010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $572.26
5020BP010CF R&D Systems
RDS5020BP010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $572.26
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Ser293-Arg408
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.1 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 12159
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
13.1 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
12159
BMP-4
10μg
Mouse