Please login to your online account to display your discounted pricing

R&D Systems Mouse Betacellulin/BTC Recombinant Protein

$415.83 - $415.83

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 12223
Molecular Weight 9.9 kDa
Name Betacellulin/BTC
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

1025CE025CF

 
r&d systems
RDS1025CE025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $415.83

1025CE025

 
r&d systems
RDS1025CE025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $415.83
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 12223
Molecular Weight 9.9 kDa
Name Betacellulin/BTC
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Mouse

Source: E. coli-derived, Asp32-Gln118
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 12223
Regulatory Status: RUO