missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse Activin C Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 16325
Molecular Weight 12.6 kDa (monomer)
Name Activin C
Purity 95%, by SDS-PAGE with silver staining.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
489AC010CF R&D Systems
RDS489AC010CF
Lyophilized from a 0.2μm filtered solution in HCl. Each for $376.73
489AC010 R&D Systems
RDS489AC010
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. Each for $376.73
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Gly237-Ser352
Purity: 95%, by SDS-PAGE with silver staining.
Molecular Weight: 12.6 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in HCl.
Entrez Gene ID: 16325
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
12.6 kDa (monomer)
95%, by SDS-PAGE with silver staining.
Yes
16325
Activin C
10μg
Mouse