Please login to your online account to display your discounted pricing

R&D Systems Mouse 4-1BB Ligand/TNFSF9 Recombinant Protein


Manufacturer: r&d systems  RDS12464L025

 View more versions of this product

Catalog No. 12464L025

  • / Each
 

Description & Specifications

Specifications

Product Type 4-1BB Ligand/TNFSF9
Host Species Mouse
Gene ID (Entrez) 21950
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight 25.7 kDa
Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Quantity 25μg
Recombinant Yes