missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Mouse 4-1BB Ligand/TNFSF9 Recombinant Protein

$428.87 - $428.87

Specifications

Gene ID (Entrez) 21950
Host Species Mouse
Molecular Weight 25.7 kDa
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
12464L025CF R&D Systems
RDS12464L025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
12464L025 R&D Systems
RDS12464L025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Arg104-Glu309, with a N-terminal Met and 10-His tag + 16 aa linker
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 25.7 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 21950
Regulatory Status: RUO

Description & Specifications

Specifications

21950
25.7 kDa
25μg
Mouse
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes