missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Wnt-7a Recombinant Protein

$506.82 - $506.82

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7476
Molecular Weight 35.5 kDa
Name Wnt-7a
Purity 70%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3008WN025 R&D Systems
RDS3008WN025
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. Each for $506.82
3008WN025CF R&D Systems
RDS3008WN025CF
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. Each for $506.82
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Leu32-Lys349
Purity: 70%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 35.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Entrez Gene ID: 7476
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
35.5 kDa
70%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
7476
Wnt-7a
25μg
Human