Please login to your online account to display your discounted pricing

R&D Systems Human Wnt-4 Recombinant Protein

$404.75 - $404.75

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 54361
Molecular Weight 39.1 kDa
Name Wnt-4
Purity 60%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

6076WN005

 
r&d systems
RDS6076WN005
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. Each for $404.75

6076WN005CF

 
r&d systems
RDS6076WN005CF
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. Each for $404.75
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 54361
Molecular Weight 39.1 kDa
Name Wnt-4
Purity 60%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 5μg
Recombinant Yes
Source Human

Source: Chinese Hamster Ovary cell line, CHO-derived, Ser23-Arg351
Purity: 60%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 39.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Entrez Gene ID: 54361
Regulatory Status: RUO