missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Wnt-11 Recombinant Protein

$456.25 - $456.25

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7481
Molecular Weight 36.6 kDa
Name Wnt-11
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
6179WN010 R&D Systems
RDS6179WN010
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. Each for $456.25
6179WN010CF R&D Systems
RDS6179WN010CF
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. Each for $456.25
Specifications

Description

Source: Chinese Hamster Ovary cell line, CHO-derived, Ile25-Lys354
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 36.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS.
Entrez Gene ID: 7481
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
36.6 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
7481
Wnt-11
10μg
Human