missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human VEGF R1/Flt-1 Fc Chimera Recombinant Protein

$467.98 - $467.98

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2321
Molecular Weight 100 kDa (monomer)
Name VEGF R1/Flt-1 Fc Chimera
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
321FL050 R&D Systems
RDS321FL050
Lyophilized from a 0.2μm filtered solution in MOPS, NaCl and CHAPS with BSA as a carrier protein. Each for $467.98
321FL050CF R&D Systems
RDS321FL050CF
Lyophilized from a 0.2μm filtered solution in MOPS, NaCl and CHAPS. Each for $467.98
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ser27-His687, with a C-terminal 6-His tag
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 100 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, NaCl and CHAPS.
Entrez Gene ID: 2321
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
100 kDa (monomer)
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
2321
VEGF R1/Flt-1 Fc Chimera
50μg
Human