Please login to your online account to display your discounted pricing

R&D Systems Human VEGF/PlGF Heterodimer Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Molecular Weight 28 kDa
Name VEGF/PlGF Heterodimer
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 5μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

297VP005

 
r&d systems
RDS297VP005
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $428.87

297VP005CF

 
r&d systems
RDS297VP005CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $428.87
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 28 kDa
Name VEGF/PlGF Heterodimer
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 5μg
Recombinant Yes
Source Human

Source: E. coli-derived, Ala27-Arg191 (VEGF) & Ala21-Arg149 (PIGF)
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 28 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Regulatory Status: RUO