Please login to your online account to display your discounted pricing

R&D Systems Human VEGF-C Recombinant Protein

$407.05 - $407.05

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7424
Molecular Weight 15.5 kDa
Name VEGF-C
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

2179VC025

 
r&d systems
RDS2179VC025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $407.05
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7424
Molecular Weight 15.5 kDa
Name VEGF-C
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Human

Source: Mouse myeloma cell line, NS0-derived, Thr103-Arg227, with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 15.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 7424
Regulatory Status: RUO