missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human VEGF 165b Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7422
Molecular Weight 19.1 kDa (monomer)
Name VEGF 165b
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
3045VE025 R&D Systems
RDS3045VE025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $428.87
3045VE025CF R&D Systems
RDS3045VE025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $428.87
Specifications

Description

Source: Spodoptera frugiperda, Sf 21 (stably transfected)-derived, Ala27-Asp191
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.1 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 7422
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
19.1 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
7422
VEGF 165b
25μg
Human