Please login to your online account to display your discounted pricing

R&D Systems Human VEGF 165 Recombinant Protein

For use in cell or tissue culture applications.

$368.77 - $5,486.88

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7422
Molecular Weight 19.2 kDa (monomer)
Name VEGF 165
Purity 97%, by SDS-PAGE with silver staining.
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

293VE050

 
r&d systems
RDS293VE050
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 50μg Each for $1,159.90

293VE010CF

 
r&d systems
RDS293VE010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose. 10μg Each for $368.77

293VE001MGC

 
r&d systems
293VE001MGCF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose. 1mg Each for $5,486.88

293VE010

 
r&d systems
RDS293VE010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 10μg Each for $368.77

293VE050CF

 
r&d systems
RDS293VE050CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose. 50μg Each for $1,159.90

29-3VE-500

 
r&d systems
293VE500
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 500μg Each for $2,922.08
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7422
Molecular Weight 19.2 kDa (monomer)
Name VEGF 165
Purity 97%, by SDS-PAGE with silver staining.
Recombinant Yes
Source Human

  • Source: Spodoptera frugiperda, Sf 21 (baculovirus)-derived, Ala27-Arg191
  • Purity: 97%, by SDS-PAGE with silver staining.
  • Molecular Weight: 19.2 kDa (monomer)
  • Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with trehalose.
  • Entrez Gene ID: 7422
  • Regulatory Status: RUO