missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human VCAM-1/CD106 Recombinant Protein

$415.83 - $1,276.19

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol.
Gene ID (Entrez) 7412
Molecular Weight 74 kDa
Name VCAM-1/CD106
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
809VR200 R&D Systems
RDS809VR200
200μg Each for $1,276.19
809VR050 R&D Systems
RDS809VR050
50μg Each for $415.83
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Phe25-Glu698, with a substitution of Arg322Gly and an N-terminal Met
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 74 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol.
Entrez Gene ID: 7412
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
7412
VCAM-1/CD106
Yes
Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol.
74 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human