missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human UCH-L1 Recombinant Protein

$158.36 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 7345
Name UCH-L1
Recombinant Yes
Source Human
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Purity Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Purity Quantity Price Quantity  
E340025 R&D Systems
RDSE340025
No Formulation Info in Bulk Inventory 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. 25μg Each for $158.36
6007CY020 R&D Systems
RDS6007CY020
Supplied as a 0.2μm filtered solution in Tris, NaCl, Glycerol and DTT. 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. 20μg Each for $416.79
Specifications

Description

Source: E. coli-derived
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 26 kDa
Formulation: Supplied as a 0.2μm filtered solution in Tris, NaCl, Glycerol and DTT.
Entrez Gene ID: 7345
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
UCH-L1
Human
7345
Yes