Please login to your online account to display your discounted pricing

R&D Systems Human Ubiquitin Agarose Recombinant Protein


Manufacturer: r&d systems  RDSU400500

 View more versions of this product

Catalog No. U400500

  • / Each
 

Description & Specifications

Specifications

Name Ubiquitin Agarose
Quantity 500μL
Recombinant Yes
Source Human
Conjugate Agarose
Formulation Supplied as a solution in Acetic Acid.
Gene ID (Entrez) 7314