Please login to your online account to display your discounted pricing

R&D Systems Human Ubiquitin Agarose Recombinant Protein


Manufacturer: r&d systems  RDSU40001M

 View more versions of this product

Catalog No. U40001M

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 7314
Name Ubiquitin Agarose
Source Human
Recombinant Yes
Quantity 1mL
Conjugate Agarose
Formulation Supplied as a solution in Acetic Acid.