missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Human Ubiquitin Agarose Recombinant Protein

$207.26 - $350.65

Specifications

Conjugate Agarose
Formulation Supplied as a solution in Acetic Acid.
Gene ID (Entrez) 7314
Name Ubiquitin Agarose
Recombinant Yes
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
U400500 R&D Systems
RDSU400500
500μL Each for $207.26
U40001M R&D Systems
RDSU40001M
1mL Each for $350.65
Specifications

Description

Source: E. coli-derived
Formulation: Supplied as a solution in Acetic Acid.
Entrez Gene ID: 7314
Regulatory Status: RUO

Description & Specifications

Specifications

Agarose
7314
Yes
Supplied as a solution in Acetic Acid.
Ubiquitin Agarose
Human