Please login to your online account to display your discounted pricing

R&D Systems Human UBE2M/Ubc12 Recombinant Protein


Manufacturer: r&d systems  RDSE2656050

 View more versions of this product

Catalog No. E2656050

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 9040
Molecular Weight 21 kDa
Name UBE2M/Ubc12
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Quantity 50μg
Recombinant Yes
Source Human
Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.