Please login to your online account to display your discounted pricing

R&D Systems Human UBE2M/Ubc12 Recombinant Protein

$172.26 - $286.85

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 9040
Molecular Weight 21 kDa
Name UBE2M/Ubc12
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2656100

 
r&d systems
RDSE2656100
100μg Each for $286.85

E2656050

 
r&d systems
RDSE2656050
50μg Each for $172.26
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 9040
Molecular Weight 21 kDa
Name UBE2M/Ubc12
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human

Source: E. coli-derived
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight: 21 kDa
Formulation: Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Entrez Gene ID: 9040
Regulatory Status: RUO