Please login to your online account to display your discounted pricing

R&D Systems Human UBE2K/E2-25K Recombinant Protein


Manufacturer: r&d systems  RDSE2603050

 View more versions of this product

Catalog No. E2603050

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in Tris, NaCl, DTT and EDTA.
Gene ID (Entrez) 3093
Molecular Weight 22 kDa
Name UBE2K/E2-25K
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Quantity 50μg
Recombinant Yes
Source Human

Source: E. coli-derived
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight: 22 kDa
Formulation: Supplied as a solution in Tris, NaCl, DTT and EDTA.
Entrez Gene ID: 3093
Regulatory Status: RUO