Please login to your online account to display your discounted pricing

R&D Systems Human UBE2K/E2-25K Recombinant Protein

$175.98 - $296.95

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in Tris, NaCl, DTT and EDTA.
Gene ID (Entrez) 3093
Molecular Weight 22 kDa
Name UBE2K/E2-25K
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2603050

 
r&d systems
RDSE2603050
50μg Each for $175.98

E2603100

 
r&d systems
RDSE2603100
100μg Each for $296.95
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in Tris, NaCl, DTT and EDTA.
Gene ID (Entrez) 3093
Molecular Weight 22 kDa
Name UBE2K/E2-25K
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human

Source: E. coli-derived
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight: 22 kDa
Formulation: Supplied as a solution in Tris, NaCl, DTT and EDTA.
Entrez Gene ID: 3093
Regulatory Status: RUO