Please login to your online account to display your discounted pricing

R&D Systems Human UBE2K/E2-25K Recombinant Protein


Manufacturer: r&d systems  RDSE2603100

 View more versions of this product

Catalog No. E2603100

  • / Each
 

Description & Specifications

Specifications

Source Human
Gene ID (Entrez) 3093
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Formulation Supplied as a solution in Tris, NaCl, DTT and EDTA.
Quantity 100μg
Recombinant Yes
Conjugate Unlabeled
Molecular Weight 22 kDa
Name UBE2K/E2-25K