Please login to your online account to display your discounted pricing

R&D Systems Human UbcH5c/UBE2D3 Recombinant Protein


Manufacturer: r&d systems  RDSE2627050

 View more versions of this product

Catalog No. E2627050

  • / Each
 

Description & Specifications

Specifications

Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Name UbcH5c/UBE2D3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight 17 kDa
Source Human
Quantity 50μg
Conjugate Unlabeled
Gene ID (Entrez) 7323
Recombinant Yes