Please login to your online account to display your discounted pricing

R&D Systems Human UbcH5c/UBE2D3 Recombinant Protein

$292.21 - $292.21

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 7323
Molecular Weight 17 kDa
Name UbcH5c/UBE2D3
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2627100

 
r&d systems
RDSE2627100
100μg Each for $292.21
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 7323
Molecular Weight 17 kDa
Name UbcH5c/UBE2D3
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human

Source: E. coli-derived
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight: 17 kDa
Formulation: Supplied as a solution in HEPES, NaCl, DTT and Glycerol.
Entrez Gene ID: 7323
Regulatory Status: RUO