Please login to your online account to display your discounted pricing

R&D Systems Human UbcH5b/UBE2D2 Recombinant Protein


Manufacturer: r&d systems  RDSE2622050

 View more versions of this product

Catalog No. E2622050

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 17 kDa
Name UbcH5b/UBE2D2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Quantity 50μg
Recombinant Yes
Source Human
Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, Glycerol.
Gene ID (Entrez) 7322