Please login to your online account to display your discounted pricing

R&D Systems Human UbcH5b/UBE2D2 Recombinant Protein

$186.40 - $298.51

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, Glycerol.
Gene ID (Entrez) 7322
Molecular Weight 17 kDa
Name UbcH5b/UBE2D2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

E2622050

 
r&d systems
RDSE2622050
50μg Each for $186.40

E2622100

 
r&d systems
RDSE2622100
100μg Each for $298.51
Description & Specifications

Specifications

Conjugate Unlabeled
Formulation Supplied as a solution in HEPES, NaCl, Glycerol.
Gene ID (Entrez) 7322
Molecular Weight 17 kDa
Name UbcH5b/UBE2D2
Purity 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Recombinant Yes
Source Human

Source: E. coli-derived
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Molecular Weight: 17 kDa
Formulation: Supplied as a solution in HEPES, NaCl, Glycerol.
Entrez Gene ID: 7322
Regulatory Status: RUO